ADVH49B82A47 AChemBlock

6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-3-carbonitrile

  • Molecular Weight : 269.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-indazole-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85270
Availability details for
Quantity
  • Availability
  • Add to Cart