ADVH49B82A48 AChemBlock

tert-butyl (1R,5S)-6-fluoro-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • Molecular Weight : 201.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-5s-6-fluoro-3-azabicyclo-3-1-0-hexane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84817
Availability details for
Quantity
  • Availability
  • Add to Cart