ADVH49B82A4A AChemBlock

6-(bromomethyl)-2,2-difluoro-spiro[2.5]octane

  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromomethyl-2-2-difluoro-spiro-2-5-octane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85273
Availability details for
Quantity
  • Availability
  • Add to Cart