ADVH49B82A4B AChemBlock

6-(benzyloxy)-8-(2-chloroacetyl)-2H-benzo[b][1,4]oxazin-3(4H)-one

  • Molecular Weight : 331.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/6-benzyloxy-8-2-chloroacetyl-2h-benzo-b-1-4-oxazin-3-4h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86466
Availability details for
Quantity
  • Availability
  • Add to Cart