ADVH49B82A4D AChemBlock

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[2,1-f][1,2,4]triazine

  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrrolo-2-1-f-1-2-4-triazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85274
Availability details for
Quantity
  • Availability
  • Add to Cart