ADVH49B82A56 AChemBlock

5-oxaspiro[3.4]octan-8-amine

  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-oxaspiro-3-4-octan-8-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85286
Availability details for
Quantity
  • Availability
  • Add to Cart