ADVH49B82A58 AChemBlock

tert-butyl 2-{[(tert-butyldiphenylsilyl)oxy]methyl}-4-(hydroxymethyl)piperidine-1-carboxylate

  • Molecular Weight : 483.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-tert-butyldiphenylsilyl-oxy-methyl-4-hydroxymethyl-piperidine-1-carboxylate-307987.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84677
Availability details for
Quantity
  • Availability
  • Add to Cart