ADVH49B82A5D AChemBlock

5-methoxy-6,7-dinitro-indoline

  • Molecular Weight : 239.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-6-7-dinitro-indoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85293
Availability details for
Quantity
  • Availability
  • Add to Cart