ADVH49B82A61 AChemBlock

5-fluoroindolin-7-amine dihydrochloride

  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoroindolin-7-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85297
Availability details for
Quantity
  • Availability
  • Add to Cart