ADVH49B82A69 AChemBlock

5-fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

  • Molecular Weight : 224.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-2-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85302
Availability details for
Quantity
  • Availability
  • Add to Cart