ADVH49B82A6E AChemBlock

5-fluoro-1H,2H,3H-pyrrolo[2,3-c]pyridin-2-one

  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-1h-2h-3h-pyrrolo-2-3-c-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85306
Availability details for
Quantity
  • Availability
  • Add to Cart