ADVH49B82A6F AChemBlock

5-fluoro-1,2,3,4-tetrahydroquinolin-8-amine hydrochloride

  • Molecular Weight : 202.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-1-2-3-4-tetrahydroquinolin-8-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85307
Availability details for
Quantity
  • Availability
  • Add to Cart