ADVH49B82A70 AChemBlock

tert-butyl 2-{[(4-methylbenzenesulfonyl)oxy]methyl}-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-5-carboxylate

  • Molecular Weight : 407.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-4-methylbenzenesulfonyl-oxy-methyl-4h-5h-6h-7h-pyrazolo-1-5-a-pyrazine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84678
Availability details for
Quantity
  • Availability
  • Add to Cart