ADVH49B82A71 AChemBlock

spiro[3.5]nonan-9-amine

  • Molecular Weight : 139.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/spiro-3-5-nonan-9-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84829
Availability details for
Quantity
  • Availability
  • Add to Cart