ADVH49B82A74 AChemBlock

5-chloro-3-methyl-imidazo[1,5-c]pyrimidine

  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-methyl-imidazo-1-5-c-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85312
Availability details for
Quantity
  • Availability
  • Add to Cart