ADVH49B82A79 AChemBlock

5-bromooxan-3-amine

  • Molecular Weight : 180.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromooxan-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85316
Availability details for
Quantity
  • Availability
  • Add to Cart