ADVH49B82A7C AChemBlock

5-bromo-7-methyl-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-7-methyl-pyrazolo-1-5-a-pyrimidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85319
Availability details for
Quantity
  • Availability
  • Add to Cart