ADVH49B82A7F AChemBlock

5-bromo-6-chloro-3-iodo-1H-indole

  • Molecular Weight : 356.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-chloro-3-iodo-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85321
Availability details for
Quantity
  • Availability
  • Add to Cart