ADVH49B82A80 AChemBlock

5-bromo-6-chloro-1H-pyrazolo[4,3-b]pyridine

  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-chloro-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85322
Availability details for
Quantity
  • Availability
  • Add to Cart