ADVH49B82A83 AChemBlock

5-bromo-4-ethyl-pyridine-2-carbonitrile

  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-ethyl-pyridine-2-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85324
Availability details for
Quantity
  • Availability
  • Add to Cart