ADVH49B82A84 AChemBlock

5-bromo-4-cyclopropyl-N,N-dimethyl-pyrazole-1-sulfonamide

  • Molecular Weight : 294.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-cyclopropyl-n-n-dimethyl-pyrazole-1-sulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85325
Availability details for
Quantity
  • Availability
  • Add to Cart