ADVH49B82A85 AChemBlock

5-bromo-4-(1,1-difluoroethyl)-1H-pyrimidin-6-one

  • Molecular Weight : 239.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-1-1-difluoroethyl-1h-pyrimidin-6-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85326
Availability details for
Quantity
  • Availability
  • Add to Cart