ADVH49B82A86 AChemBlock

5-bromo-3-methyl-1-(oxan-2-yl)-1H-pyrazole

  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-methyl-1-oxan-2-yl-1h-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85327
Availability details for
Quantity
  • Availability
  • Add to Cart