ADVH49B82A87 AChemBlock

5-Bromo-3-methoxy-pyrazine-2-carbaldehyde

  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-methoxy-pyrazine-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86646
Availability details for
Quantity
  • Availability
  • Add to Cart