ADVH49B82A88 AChemBlock

pyrrolo[2,1-f][1,2,4]triazine-4-carboxylic acid

  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/pyrrolo-2-1-f-1-2-4-triazine-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84837
Availability details for
Quantity
  • Availability
  • Add to Cart