ADVH49B82A89 AChemBlock

potassium trifluoro-(3-methoxycarbonyl-1-bicyclo[1.1.1]pentanyl)boranuide

  • Molecular Weight : 232.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/potassium-trifluoro-3-methoxycarbonyl-1-bicyclo-1-1-1-pentanyl-boranuide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84838
Availability details for
Quantity
  • Availability
  • Add to Cart