ADVH49B82A8B AChemBlock

potassium bicyclo[1.1.1]pentan-1-yltrifluoroborate

  • Molecular Weight : 174.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/potassium-bicyclo-1-1-1-pentan-1-yltrifluoroborate-308150.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84840
Availability details for
Quantity
  • Availability
  • Add to Cart