ADVH49B82A91 AChemBlock

potassium [(4,4-difluorocyclohexyl)methyl]trifluoroboranuide

  • Molecular Weight : 240.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/potassium-4-4-difluorocyclohexyl-methyl-trifluoroboranuide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84841
Availability details for
Quantity
  • Availability
  • Add to Cart