ADVH49B82A92 AChemBlock

oxane-3,5-diol

  • Molecular Weight : 118.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/oxane-3-5-diol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84842
Availability details for
Quantity
  • Availability
  • Add to Cart