ADVH49B82A94 AChemBlock

O-Methyl-L-tyrosine methyl ester hydrochloride

  • Molecular Weight : 245.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/o-methyl-l-tyrosine-methyl-ester-hydrochloride-293709.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70399
Availability details for
Quantity
  • Availability
  • Add to Cart