ADVH49B82A97 AChemBlock

5-bromo-1,1-dimethyl-isoindoline hydrochloride

  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-1-dimethyl-isoindoline-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85336
Availability details for
Quantity
  • Availability
  • Add to Cart