ADVH49B82A99 AChemBlock

5-bromo-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile

  • Molecular Weight : 223.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-2-4-triazolo-1-5-a-pyridine-8-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85338
Availability details for
Quantity
  • Availability
  • Add to Cart