ADVH49B82A9C AChemBlock

tert-butyl 2-[(thietan-3-yl)amino]propanoate

  • Molecular Weight : 217.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-thietan-3-yl-amino-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84681
Availability details for
Quantity
  • Availability
  • Add to Cart