ADVH49B82A9E AChemBlock

5-{spiro[3.3]heptan-2-yl}-1H-pyrazol-3-amine

  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-spiro-3-3-heptan-2-yl-1h-pyrazol-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85341
Availability details for
Quantity
  • Availability
  • Add to Cart