ADVH49B82AA1 AChemBlock

5-[(propan-2-yloxy)methyl]-1H-pyrazole-3-carboxylic acid

  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-propan-2-yloxy-methyl-1h-pyrazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85343
Availability details for
Quantity
  • Availability
  • Add to Cart