ADVH49B82AA4 AChemBlock

5-[(2-methylpropoxy)methyl]-1H-pyrazole-3-carboxylic acid

  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-2-methylpropoxy-methyl-1h-pyrazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85346
Availability details for
Quantity
  • Availability
  • Add to Cart