ADVH49B82AA6 AChemBlock

5,6-dichloro-3-iodo-1H-indole

  • Molecular Weight : 311.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-6-dichloro-3-iodo-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85348
Availability details for
Quantity
  • Availability
  • Add to Cart