ADVH49B82AA8 AChemBlock

tert-butyl 3-iodo-2-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-4-carboxylate

  • Molecular Weight : 363.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-iodo-2-methyl-4h-5h-6h-7h-pyrazolo-1-5-a-pyrimidine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84637
Availability details for
Quantity
  • Availability
  • Add to Cart