ADVH49B82AAA AChemBlock

O2-tert-butyl O5-methyl 2-azabicyclo[2.2.2]octane-2,5-dicarboxylate

  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o2-tert-butyl-o5-methyl-2-azabicyclo-2-2-2-octane-2-5-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84851
Availability details for
Quantity
  • Availability
  • Add to Cart