ADVH49B82AB2 AChemBlock

5-(bromomethyl)-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-pyrazole

  • Molecular Weight : 291.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromomethyl-1-2-trimethylsilyl-ethoxy-methyl-1h-pyrazole-308669.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85359
Availability details for
Quantity
  • Availability
  • Add to Cart