ADVH49B82AB4 AChemBlock

5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-1H-quinolin-2-one

  • Molecular Weight : 273.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-3-4-dihydro-1h-quinolin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85361
Availability details for
Quantity
  • Availability
  • Add to Cart