ADVH49B82AB6 AChemBlock

4-methyl-octahydro-1H-pyrrolo[3,2-b]pyridine dihydrochloride

  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-methyl-octahydro-1h-pyrrolo-3-2-b-pyridine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85363
Availability details for
Quantity
  • Availability
  • Add to Cart