ADVH49B82ABD AChemBlock

O1-tert-butyl O2-methyl 4-(hydroxymethyl)piperidine-1,2-dicarboxylate

  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o1-tert-butyl-o2-methyl-4-hydroxymethyl-piperidine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84857
Availability details for
Quantity
  • Availability
  • Add to Cart