ADVH49B82ABE AChemBlock

O1-tert-butyl O2-ethyl (2S,5R)-5-aminopiperidine-1,2-dicarboxylate

  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o1-tert-butyl-o2-ethyl-2s-5r-5-aminopiperidine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84858
Availability details for
Quantity
  • Availability
  • Add to Cart