ADVH49B82AC8 AChemBlock

4-Fluoro-5-methyl-biphenyl-3-carbaldehyde

  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-5-methyl-biphenyl-3-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86867
Availability details for
Quantity
  • Availability
  • Add to Cart