ADVH49B82ACD AChemBlock

O1-benzyl O2-methyl (2R,4R)-4-hydroxy-2-methyl-pyrrolidine-1,2-dicarboxylate

  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o1-benzyl-o2-methyl-2r-4r-4-hydroxy-2-methyl-pyrrolidine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84864
Availability details for
Quantity
  • Availability
  • Add to Cart