ADVH49B82ACF AChemBlock

N-methyl-N-[(3S)-3-piperidyl]acetamide hydrochloride

  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-n-3s-3-piperidyl-acetamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84866
Availability details for
Quantity
  • Availability
  • Add to Cart