ADVH49B82AE9 AChemBlock

4-bromopyridine-3-carboxylic acid hydrobromide

  • Molecular Weight : 282.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromopyridine-3-carboxylic-acid-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71903
Availability details for
Quantity
  • Availability
  • Add to Cart