ADVH49B82AEE AChemBlock

methyl trans-3-fluorocyclopentane-1-carboxylate

  • Molecular Weight : 146.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-trans-3-fluorocyclopentane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84873
Availability details for
Quantity
  • Availability
  • Add to Cart