ADVH49B82AF0 AChemBlock

methyl trans-1-amino-3-methoxycyclobutanecarboxylate hydrochloride

  • Molecular Weight : 195.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-trans-1-amino-3-methoxycyclobutanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84875
Availability details for
Quantity
  • Availability
  • Add to Cart